eBioscience™ Streptavidin FITC Conjugate
Invitrogen™

eBioscience™ Streptavidin FITC Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Have Questions?
產品號碼Quantity
11-4317-87250 μg
產品號碼 11-4317-87
價格 (TWD)
3,656.00
線上優惠
Ends: 30-Sep-2026
4,570.00
您節省 914.00 (20%)
Each
Quantity:
250 μg
價格 (TWD)
3,656.00
線上優惠
Ends: 30-Sep-2026
4,570.00
您節省 914.00 (20%)
Each

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Blue Laser

Emit

520 nm

Excite

488 nm

Reported Application

Flow Cytometric Analysis, Immunocytochemistry, Immunohistochemical Staining of Frozen Tissue Sections

For Research Use Only. Not for use in diagnostic procedures.
規格
Concentration0.5 mg/mL
Label or DyeFITC (Fluorescein)
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity250 μg
Target MoleculeBiotin
ConjugateFITC (Fluorescein)
FormLiquid
Product LineeBioscience
Unit SizeEach