2-Chloroquinoline, 98%
2-Chloroquinoline, 98%
2-Chloroquinoline, 98%
Thermo Scientific Chemicals

2-Chloroquinoline, 98%

CAS: 612-62-4 | C9H6ClN | 163.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 110040250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS612-62-4
IUPAC Name2-chloroquinoline
Molecular FormulaC9H6ClN
InChI KeyOFUFXTHGZWIDDB-UHFFFAOYSA-N
SMILESClC1=CC=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Form)or fused mass
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only