AlbuMAX™ I Lipid-Rich BSA, 25 g - FAQs
View additional product information for AlbuMAX™ I Lipid-Rich BSA - FAQs (11020039, 11020021)
1 product FAQs found
What fatty acids are in my specific lot of AlbuMAX I? What are their approximate concentrations?
Below are the concentrations of free fatty acids obtained from the HPLC lipid profiles of an older lot of AlbuMAX I:
Alpha-linolenic acid: 0.59 mg FFA/g protein
Linoleic acid: 0.65 mg FFA/g protein
Oleic acid: 2.12 mg FFA/g protein
Stearic acid: 2.09 mg FFA/g protein
Palmitic acid: 1.90 mg FFA/g protein
Note: The individual lipid concentrations may vary between lots, but the values above can be used as a good approximation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.