Product Image
Gibco™

AlbuMAX™ I Lipid-Rich BSA

AlbuMAX™ I Lipid-Rich BSA
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
110200391 kg
1102002125 g
Catalog number 11020039
Price (USD)
4,160.00
Each
Quantity:
1 kg
Price (USD)
4,160.00
Each

Lipid-rich bovine serum albumin for cell culture (1). It is chromatographically purified and has an IgG content ≤ 0.1%.

Origin: New Zealand

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Product LineAlbuMAX, Gibco
Quantity1 kg
Shipping ConditionRoom Temperature
Product TypeBSA
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

What fatty acids are in my specific lot of AlbuMAX I? What are their approximate concentrations?

Below are the concentrations of free fatty acids obtained from the HPLC lipid profiles of an older lot of AlbuMAX I:

Alpha-linolenic acid: 0.59 mg FFA/g protein
Linoleic acid: 0.65 mg FFA/g protein
Oleic acid: 2.12 mg FFA/g protein
Stearic acid: 2.09 mg FFA/g protein
Palmitic acid: 1.90 mg FFA/g protein
Note: The individual lipid concentrations may vary between lots, but the values above can be used as a good approximation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.