Interested in a bulk or custom version of this product? Request pricing.

TFRC Recombinant Human Protein, N-His Tag

Catalog number: 11020H07H25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Transferrin Receptor (p90 CD71) (TFRC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TFRC (NP_003225.2) (Cys 89-Phe 760) fused to a polyhistidine tag at the N-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized human TFRC at 10 µg/ml (100 µl/well) can bind human Transferrin. The EC50 of human Transferrin is 5.6 ng/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by SINO Biological.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins, Other Proteins
Research Category: Biomarker-Related Pathway(s), Bone Research, Other Research
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CD71, TFR, TFR1, TRFR
Gene Symbol: TFRC
Molecular Weight: 77.4 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Name: transferrin receptor (p90, CD71)
Protein Subtype: Transferrin
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols