Interested in a bulk or custom version of this product? Request pricing.

PARP1 Recombinant Human Protein, His Tag

Catalog number: 11040H08B5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Poly (Adp-Ribose) Polymerase 1 (PARP1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from The amino acids corresponding to the full length of human PARP1 (NP_001609.2) (Met 1-Trp 1014) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized human PARP1 at 10 µg/ml (100 µl/well) can bind to biotinylated human HSP70. The EC50 of biotinylated human HSP70 is 0.035 µg/ml.

Formulation: 50 mM Tris, 100 mM NaCl, pH 8, 2 mM EDTA, 2 mM PMSF, 0.5 mM TCEP, 10% glycerol.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: PARP1, RP11-125A15.2, ADPRT, ADPRT1, PARP, PARP-1, PPOL, pADPRT-1
Gene Symbol: PARP1
Molecular Weight: 114.5 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Name: poly (ADP-ribose) polymerase 1
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: DNA Replication & Repair
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols