Thermo Scientific Chemicals

Citraconic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 110431000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Citraconic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 110431000
100 g, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
498-23-7
IUPAC Name
(2Z)-2-methylbut-2-enedioic acid
Molecular Formula
C5H6O4
InChI Key
HNEGQIOMVPPMNR-IHWYPQMZSA-N
SMILES
C\C(=C\C(O)=O)C(O)=O
Appearance (Form)
Crystalline powder
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Titration with NaOH
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text