o-Cresol, 99%
o-Cresol, 99%
o-Cresol, 99%
Thermo Scientific Chemicals

o-Cresol, 99%

CAS: 95-48-7 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 110550025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS95-48-7
IUPAC Name2-methylphenol
Molecular FormulaC7H8O
InChI KeyQWVGKYWNOKOFNN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid or liquid
GC>=98.5 %
AppearanceMay darken on storage
Appearance (Color)Colorless to amber
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only