Shandon™ Mobile Carts, Bariatric - FAQs
Shandon™ Mobile Carts, Bariatric - FAQs
View additional product information for Shandon™ Mobile Carts, Bariatric - FAQs (11058)
1 product FAQs found
How much serum is in Opti-MEM I Medium?
There is no serum in Opti-MEM I Medium. The term "reduced serum" means that one can reduce the amount of serum that is used with classical cell culture media. When working with Opti-MEM I medium, most cells that are routinely cultured in serum-supplemented medium may be transferred directly into Opti-MEM I medium with a minimum of 50% reduction in serum.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.