Interested in a bulk or custom version of this product? Request pricing.

Neuraminidase (NA) Recombinant Viral Protein, N-His Tag

Catalog number: 11058V07B25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NA Recombinant Influenza A Virus Protein subtype H1N1 (A/California/04 /2009) (NEURAMINIDASE), recombinant viral protein is supplied as a frozen protein. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of influenza A virus (A/California/04/2009 (H1N1)) neuraminidase (ACP41107.1) (His 36 -Lys 469) fused to a polyhistidine tag at the N-terminus.

Activity: Measured by its ability to cleave a fluorogenic substrate, 2'-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid. The specific activity is 800-1200 pmoles/min/µg.

Formulation: 50 mM Tris-Citrate, 0.3 M NaCl, pH6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Virology
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen, Lyophilized
Gene Aliases: NA
Gene Symbol: NA, Neuraminidase
Molecular Weight: 50 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: NA Recombinant Influenza A Virus Protein subtype H1N1 (A/California/04 /2009), Neuraminidase
Protein Subtype: Viral Proteins
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Species: Influenza
Tag Position: N-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols