Interested in a bulk or custom version of this product? Request pricing.

HA Recombinant Influenza A Virus Protein, subtype H7N7 (A/Netherlands/219/03), (His Tag)

Catalog number: 11082V08B50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Hemagglutinin, Influenza A virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the native extracellular domain (Met 8-Phe 544) of influenza A virus, subtype H7N7 (A/Netherlands/219/03), HA (AAR02640.1) (Met 8-Phe 544), fused to a polyhistidine tag at the C-terminus.

Activity: Measured in hemagglutination test using 1% chicken erythrocyte. The HA titer is 0.1-0.5 µg/mlfor 1% chicken red blood cells.

Formulation: Lyophilized in 20 MM Tris, pH 7.4, 500 mM NaCl, (web PBS ?)10% glycerol, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Microbiology, Virology
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Lyophilized
Gene Aliases: HA
Gene Symbol: HA
Molecular Weight: 59 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Family: Other Proteins
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: Hemagglutinin
Protein Subtype: Viral Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Species: Influenza
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols