Cyanuric chloride, 99%
Cyanuric chloride, 99%
Cyanuric chloride, 99%
Thermo Scientific Chemicals

Cyanuric chloride, 99%

CAS: 108-77-0 | C3Cl3N3 | 184.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 110890051
Price (TWD)
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS108-77-0
IUPAC Nametrichloro-1,3,5-triazine
Molecular FormulaC3Cl3N3
InChI KeyMGNCLNQXLYJVJD-UHFFFAOYSA-N
SMILESClC1=NC(Cl)=NC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Anticaking agentMay contain 0% to 0.5% Silica as an additive
Infrared spectrumConforms
Melting point145°C to 148°C
Titration98.5 to 101.5 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only