Cyclobutanecarboxylic acid, 98%
Cyclobutanecarboxylic acid, 98%
Cyclobutanecarboxylic acid, 98%
Thermo Scientific Chemicals

Cyclobutanecarboxylic acid, 98%

CAS: 3721-95-7 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 110900050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3721-95-7
IUPAC Namecyclobutanecarboxylic acid
Molecular FormulaC5H8O2
InChI KeyTXWOGHSRPAYOML-UHFFFAOYSA-N
SMILESOC(=O)C1CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4410 to 1.4450 (20°C, 589 nm)
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only