Cyclobutanecarboxylic acid chloride, 99%
Cyclobutanecarboxylic acid chloride, 99%
Cyclobutanecarboxylic acid chloride, 99%
Thermo Scientific Chemicals

Cyclobutanecarboxylic acid chloride, 99%

CAS: 5006-22-4 | C5H7ClO | 118.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 110910010
Price (USD)
26.65
Online exclusive
29.50
Save 2.85
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
26.65
Online exclusive
29.50
Save 2.85
Each
Chemical Identifiers
CAS5006-22-4
IUPAC Namecyclobutanecarbonyl chloride
Molecular FormulaC5H7ClO
InChI KeyJFWMYCVMQSLLOO-UHFFFAOYSA-N
SMILESClC(=O)C1CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration RCOCl98.5 to 101.5 %
Refractive index1.4540 to 1.4560 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only