Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Cover, 100 gallon - FAQs
View additional product information for Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Cover - FAQs (11100-0080, 11100-0005, 11100-0015, 11100-0007, 11100-0100, 11100-0055, 11100-0200, 11100-0030, 11100-0150, 11100-0010)
1 product FAQs found
How accurate are the molded or printed graduations on your bottles, beakers, carboys and tanks?
The accuracy of these graduations cannot be guaranteed. We suggest that they not be relied upon for measurement, but rather to be used for reference only.