Cycloheptane, 99%
Cycloheptane, 99%
Cycloheptane, 99%
Thermo Scientific Chemicals

Cycloheptane, 99%

CAS: 291-64-5 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 111011000
Price (TWD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS291-64-5
IUPAC Namecycloheptane
Molecular FormulaC7H14
InChI KeyDMEGYFMYUHOHGS-UHFFFAOYSA-N
SMILESC1CCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only