TraceCERT™ o-Xylene Solution, 5,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

TraceCERT™ o-Xylene Solution, 5,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Have Questions?
Catalog NumberQuantity
11102-00551 mL
Catalog number 11102-0055
HSN 39233090
Price (INR)
28,803.00
Case
Quantity:
1 mL
Request bulk or custom format
Price (INR)
28,803.00
Case
Chemical Identifiers
CAS95-47-6
Specifications
PackagingAmpule
Product LineTraceCERT
CAS95-47-6
RTECS NumberZE2450000
Flash Point31°C
View more
Environmental
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC