Cycloheptyl bromide, 97%
Cycloheptyl bromide, 97%
Cycloheptyl bromide, 97%
Thermo Scientific Chemicals

Cycloheptyl bromide, 97%

CAS: 2404-35-5 | C7H13Br | 177.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 111081000
Price (USD)
379.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
379.00
Each
Chemical Identifiers
CAS2404-35-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Clear light yellow
Refractive index1.5035 to 1.5055 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only