Cyclohexane, 99+%, pure
Cyclohexane, 99+%, pure
Cyclohexane, 99+%, pure
Thermo Scientific Chemicals

Cyclohexane, 99+%, pure

CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 111110010
Price (USD)
87.10
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
87.10
Each
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.005 %
Water=<0.05 % (K.F.)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only