MEM Vitamin Solution (100X) - FAQs
View additional product information for MEM Vitamin Solution (100X) - FAQs (11120037)
2 product FAQs found
What is the best way to handle the MEM Vitamin Solution (100X)? It comes frozen, so should I thaw it, then aliquot it and freeze the aliquots for later use?
If you're going to be using small batches of the vitamin solution at a time, it would be best to aliquot it out and store at -20 degrees C. The stability of the MEM Vitamin Solution is one year at -20 degrees C, while it is only 2 weeks at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the solvent used for MEM Vitamin Solution (100X)?
The solvent is distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.