Brinster's, no Glutamine, no phenol red
Brinster's, no Glutamine, no phenol red
Brinster's, no Glutamine, no phenol red
Gibco™

Brinster's, no Glutamine, no phenol red

Brinster's medium for ovum culture (BMOC) is a simple chemically defined media containing bovine serum albumin (BSA) developed by Ralph자세히 알아보기
Have Questions?
카탈로그 번호수량
11126034100 mL
카탈로그 번호 11126034
제품 가격(KRW)
85,000
온라인 행사
Ends: 30-Sep-2026
94,000
할인액 9,000 (10%)
Each
수량:
100 mL
제품 가격(KRW)
85,000
온라인 행사
Ends: 30-Sep-2026
94,000
할인액 9,000 (10%)
Each

Brinster's medium for ovum culture (BMOC) is a simple chemically defined media containing bovine serum albumin (BSA) developed by Ralph Brinster for in vitro development of mouse oocytes. This medium is a variation of the original formulation as it does not contain antibiotics. Brinster's medium has also been used for culture of other mammalian embryos during the early stages of development.

This Brinster's medium is manufactured as follows:
WithWithout
• Bovine Serum Albumin• L-Glutamine
• Sodium pyruvate• HEPES
 • Phenol Red


The complete formulation is available.

Gibco™ Brinster's medium is unique from other media as it contains no amino acids or vitamins. Brinster's medium is supplemented with bovine serum albumin required for supporting oocyte maturation and the early stages of mammalian embryo development and pyruvate as an essential energy source.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Brinster's medium uses a sodium bicarbonate buffer system (2.1 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use Only. Not for use in diagnostic procedures.
사양
세포 유형Mouse Oocytes
농도1 X
제조 품질cGMP-compliant under the ISO 13485 standard
제품라인Gibco
제품 유형Brinster's Medium
수량100 mL
유통 기한12 Months From Date of Manufacture
배송 조건Room Temperature
분류Animal Origin
형태Liquid
멸균Sterile-filtered
첨가제 포함Low Glucose, Sodium Pyruvate, Bovine Serum Albumin (BSA)
첨가제 없음No Glutamine, No HEPES, No Phenol Red
Unit SizeEach
구성 및 보관
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

자주 묻는 질문(FAQ)

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

My cells are not adhering to the culture vessel. What should I do?

This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.