Cyclohexylacetic acid, 98+%
Cyclohexylacetic acid, 98+%
Cyclohexylacetic acid, 98+%
Thermo Scientific Chemicals

Cyclohexylacetic acid, 98+%

CAS: 5292-21-7 | C8H14O2 | 142.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 111271000
Price (EUR)
409,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
409,00
Each
Chemical Identifiers
CAS5292-21-7
IUPAC Name2-cyclohexylacetic acid
Molecular FormulaC8H14O2
InChI KeyLJOODBDWMQKMFB-UHFFFAOYSA-N
SMILESOC(=O)CC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point27°C to 31°C
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Appearance (Form)Low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only