Cyclohexylmethyl bromide, 96%
Cyclohexylmethyl bromide, 96%
Cyclohexylmethyl bromide, 96%
Thermo Scientific Chemicals

Cyclohexylmethyl bromide, 96%

CAS: 2550-36-9 | C7H13Br | 177.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 111350250
Price (USD)
71.65
Online exclusive
79.40
Save 7.75
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
71.65
Online exclusive
79.40
Save 7.75
Each
Chemical Identifiers
CAS2550-36-9
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4910 to 1.4930 (20°C, 589 nm)
Appearance (Color)Clear light yellow
GC>=95 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only