Interested in a bulk or custom version of this product? Request pricing.

S100A8 Recombinant Human Protein, N-His Tag

Catalog number: 11138H07E25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

S100 Calcium Binding Protein A8 (S100A8), recombinant human protein is supplied in frozen format. It is suitable for use in performing cell-based assays and protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human S100A8 (NP_002955.2) (Leu 2-Glu 93) fused to a polyhistidine tag at the N-terminus.

Activity: Measured by its ability to bind recombinant human S100A9 in a functional ELISA. Measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells. The ED50 for this effect is typically 10-30 µg/ml.

Formulation: 20 mM Tris, 10% glycerol, pH 8.5.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Frozen
Gene Aliases: 60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8
Gene Symbol: S100A8
Molecular Weight: 12.2 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Name: S100 calcium binding protein A8
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: Signal Transduction
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols