MEM Non-Essential Amino Acids Solution (100X), 100 mL - FAQs
View additional product information for MEM Non-Essential Amino Acids Solution (100X) - FAQs (11140050, 11140076)
3 product FAQs found
Where can I find the formulation of MEM Non-Essential Amino Acids Solution (100X) (Cat. No. 11140050)?
The formulation of MEM Non-Essential Amino Acids Solution (100X) can be found at this link: https://www.thermofisher.com/us/en/home/technical-resources/media-formulation.165.html
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the solvent used for MEM Non-Essential Amino Acids Solution (100X)?
The solvent is distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is Gibco MEM Non-Essential Amino Acids?
MEM Non-Essential Amino Acids are used as a supplement for cell culture medium, to increase cell growth and viability. MEM Non-Essential Amino Acids contains the same non-essential amino acids found in the standard Minimum Essential Medium (MEM) at a strength of 100X. The complete formulation is available here (http://www.thermofisher.com/us/en/home/technical-resources/media-formulation.165.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.