Cyclooctanone, 98%
Cyclooctanone, 98%
Cyclooctanone, 98%
Thermo Scientific Chemicals

Cyclooctanone, 98%

CAS: 502-49-8 | C8H14O | 126.2 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 111445000
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS502-49-8
IUPAC Namecyclooctanone
Molecular FormulaC8H14O
InChI KeyIIRFCWANHMSDCG-UHFFFAOYSA-N
SMILESO=C1CCCCCCC1
さらに表示
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Colorless to white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only