Interested in a bulk or custom version of this product? Request pricing.

CTLA4 (CTLA-4) Recombinant Human Protein, hIgG1-Fc.His Tag

Catalog number: 11159H03H250

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Cytotoxic T-lymphocyte-associated protein 4 (CTLA4), recombinant human protein is supplied as a lyophilized powder.

This recombinant protein was expressed from a DNA sequence encoding the human CTLA4 (NP_005205.2) extracellular domain (Met 1-Phe 162) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Immunology, Virology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CTLA4, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, ICOS, IDDM12
Gene ID: 1493
Gene Symbol: CTLA4
Molecular Weight: 41.6 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Form: Recombinant, Truncated
Protein Name: Cytotoxic T-lymphocyte-associated protein 4
Protein Tag or Fusion: IgG1 Fc His
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols