Cyclopentyl bromide, 98%
Cyclopentyl bromide, 98%
Cyclopentyl bromide, 98%
Acros Organics

Cyclopentyl bromide, 98%

CAS: 137-43-9 | C5H9Br | 149.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 111600500
also known as 11160
HSN 29035900
Price (INR)
5,969.00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,969.00
Each
Chemical Identifiers
CAS137-43-9
IUPAC Namebromocyclopentane
Molecular FormulaC5H9Br
InChI KeyBRTFVKHPEHKBQF-UHFFFAOYSA-N
SMILESBrC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4870 to 1.4910 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only