3-Cyclopentylpropionic acid, 98%
3-Cyclopentylpropionic acid, 98%
3-Cyclopentylpropionic acid, 98%
Thermo Scientific Chemicals

3-Cyclopentylpropionic acid, 98%

CAS: 140-77-2 | C8H14O2 | 142.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 111610050
Price (USD)
20.65
Special offer
Online exclusive
Ends: 28-Dec-2026
23.80
Save 3.15
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
20.65
Special offer
Online exclusive
Ends: 28-Dec-2026
23.80
Save 3.15
Each
Chemical Identifiers
CAS140-77-2
IUPAC Name3-cyclopentylpropanoic acid
Molecular FormulaC8H14O2
InChI KeyZRPLANDPDWYOMZ-UHFFFAOYSA-N
SMILESOC(=O)CCC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4560 to 1.4580 (20°C, 589 nm)
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only