Cyclopropanecarboxylic acid, 98%
Cyclopropanecarboxylic acid, 98%
Cyclopropanecarboxylic acid, 98%
Thermo Scientific Chemicals

Cyclopropanecarboxylic acid, 98%

CAS: 1759-53-1 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 111621000
Price (TWD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1759-53-1
IUPAC Namecyclopropanecarboxylic acid
Molecular FormulaC4H6O2
InChI KeyYMGUBTXCNDTFJI-UHFFFAOYSA-N
SMILESOC(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only