L-Cysteine ethyl ester hydrochloride, 98%
L-Cysteine ethyl ester hydrochloride, 98%
L-Cysteine ethyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Cysteine ethyl ester hydrochloride, 98%

CAS: 868-59-7 | C5H12ClNO2S | 185.67 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 111790250
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS868-59-7
IUPAC Namehydrogen ethyl (2R)-2-amino-3-sulfanylpropanoate chloride
Molecular FormulaC5H12ClNO2S
InChI KeyJFKJWWJOCJHMGV-WCCKRBBISA-N
SMILES[H+].[Cl-].CCOC(=O)[C@@H](N)CS
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder and/or chunks
Loss on drying=<0.5 % (40°C, 5 hrs) (vacuum, P2O5)
Specific optical rotation-9° to -13° (20°C, 589 nm) (c=1, 1M HCl)
Infrared spectrumConforms
Sulfated ash=<0.1 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only