Decafluorobiphenyl, 99%
Decafluorobiphenyl, 99%
Decafluorobiphenyl, 99%
Thermo Scientific Chemicals

Decafluorobiphenyl, 99%

CAS: 434-90-2 | C12F10 | 334.12 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 111830050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS434-90-2
IUPAC Namedecafluoro-1,1'-biphenyl
Molecular FormulaC12F10
InChI KeyONUFSRWQCKNVSL-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
さらに表示
Appearance (Form)Crystalline solid
Melting point66°C to 70°C
GC>=98.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only