Decahydronaphthalene, 98%, mixture of cis and trans
Decahydronaphthalene, 98%, mixture of cis and trans
Decahydronaphthalene, 98%, mixture of cis and trans
Thermo Scientific Chemicals

Decahydronaphthalene, 98%, mixture of cis and trans

CAS: 91-17-8 | C10H18 | 138.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number 111840010
Price (USD)
115.65
Special offer
Online exclusive
Ends: 28-Dec-2026
136.00
Save 20.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
115.65
Special offer
Online exclusive
Ends: 28-Dec-2026
136.00
Save 20.35
Each
Chemical Identifiers
CAS91-17-8
IUPAC Namedecahydronaphthalene
Molecular FormulaC10H18
InChI KeyNNBZCPXTIHJBJL-UHFFFAOYSA-N
SMILESC1CCC2CCCCC2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4730 to 1.4760 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only