n-Decylamine, 99%
n-Decylamine, 99%
n-Decylamine, 99%
Thermo Scientific Chemicals

n-Decylamine, 99%

CAS: 2016-57-1 | C10H23N | 157.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 111920100
Price (EUR)
49,65
Online exclusive
55,00
Save 5,35 (10%)
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
49,65
Online exclusive
55,00
Save 5,35 (10%)
Each
Chemical Identifiers
CAS2016-57-1
IUPAC Namedecan-1-amine
Molecular FormulaC10H23N
InChI KeyMHZGKXUYDGKKIU-UHFFFAOYSA-N
SMILESCCCCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to yellow to brown
Refractive index1.4350 to 1.4370 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only