2-Deoxy-D-glucose, 98+%, Thermo Scientific Chemicals
2-Deoxy-D-glucose, 98+%, Thermo Scientific Chemicals
2-Deoxy-D-glucose, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Deoxy-D-glucose, 98+%, Thermo Scientific Chemicals

CAS: 154-17-6 | C6H12O5 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 111980050
Price (EUR)
143,65
Online exclusive
159,00
Save 15,35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
143,65
Online exclusive
159,00
Save 15,35
Each
Chemical Identifiers
CAS154-17-6
IUPAC Name(4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Molecular FormulaC6H12O5
InChI KeyPMMURAAUARKVCB-PHUJZJCSNA-N
SMILESOC[C@H]1OC(O)C[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
HPLC>=98.0 %
Loss on drying=<1 % (1 g, 105°C, 3 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only