Diacetone-D-glucose, 98+%
Diacetone-D-glucose, 98+%
Diacetone-D-glucose, 98+%
Thermo Scientific Chemicals

Diacetone-D-glucose, 98+%

CAS: 582-52-5 | C12H20O6 | 260.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 112015000
Price (USD)
615.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
615.00
Each
Chemical Identifiers
CAS582-52-5
IUPAC Name5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Molecular FormulaC12H20O6
InChI KeyKEJGAYKWRDILTF-UHFFFAOYNA-N
SMILESCC1(C)OCC(O1)C1OC2OC(C)(C)OC2C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
GC>=98.0 %
Sulfated ash=<0.2 %
Loss on drying=<0.5 %
Specific optical rotation-10° to -12° (20°C, 589 nm) (c=5, EtOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only