Diallylamine, 97%
Diallylamine, 97%
Diallylamine, 97%
Thermo Scientific Chemicals

Diallylamine, 97%

CAS: 124-02-7 | C6H11N | 97.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 112042500
Price (USD)
52.65
Online exclusive
58.50
Save 5.85
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
52.65
Online exclusive
58.50
Save 5.85
Each
Chemical Identifiers
CAS124-02-7
IUPAC Namebis(prop-2-en-1-yl)amine
Molecular FormulaC6H11N
InChI KeyDYUWTXWIYMHBQS-UHFFFAOYSA-N
SMILESC=CCNCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4395 to 1.4415 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only