Interested in a bulk or custom version of this product? Request pricing.

HPGD Recombinant Human Protein, His Tag

Catalog number: 11205H08E5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Hydroxyprostaglandin Dehydrogenase 15-(Nad) (HPGD), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human HPGD (NP_000851.2) (Met 1-Gln 266) fused to a polyhistidine tag at the C-terminus.

Activity: Measure by the production of NADH during the oxidation of PGF2-alpha. The specific activity is >1, 500 pmoles/min/µg.

Formulation: 50 mM Tris, 100 mM NaCl, 0.5 mM DTT, 0.2 µg/µl BSA, 10% glycerol, pH.7.5

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Frozen
Gene Aliases: 15-PGDH, PGDH, PGDH1, SDR36C1, HPGD
Gene Symbol: HPGD
Molecular Weight: 29.7 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Name: hydroxyprostaglandin dehydrogenase 15-(NAD)
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Research Category: Other Research
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols