EA IsoLink™ IRMS System
Thermo Scientific™
EA IsoLink™ IRMS System
Get higher throughput and precision for your C, N, S, O, and H isotope ratio measurements using the Thermo Scientific™ EA IsoLink IRMS System.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoType
11206185CNSOH IRMS System
11206175CN IRMS System
11206180CN/OH IRMS System
BRE723644OH IRMS System
Número do catálogo 11206185
Preço (BRL)
-
Type:
CNSOH IRMS System
Discover an automated, easy-to-use solution for μg- to mg-level elemental and isotope analysis with the Thermo Scientific™ EA IsoLink™ IRMS System. This all-in-one isotope ratio mass spectrometry system enables laboratories to increase sample throughput, reduce cost per sample, and provides high-precision elemental and isotopic analysis. Innovation in the EA IsoLink IRMS System provides helium saving technology and integration of temperature ramped gas chromatography delivering quick analysis times and low cost analysis alongside outstanding quality data, especially on small sample amounts. These features allow you to push boundaries for research and routine analyses in ecology, biology, archaeology, geosciences, food integrity, and forensics whilst offering built-in flexibility to quickly and simply adapt to changing analytical demands.
The EA IsoLink IRMS System uses temperature-ramped gas chromatography (GC), which reduces analysis time per sample, increases sample throughput, and produces high quality data thanks to optimal baseline separation of analyte gases and sharp peak shapes. These benefits are the prerequisites for high-performance IRMS.
Low Helium Consumption
Use the Thermo Scientific™ Intelligent Helium Management (HeM) Module to ensure low helium consumption during your analysis and significantly reduce your sample analysis costs.
EA IsoLink IRMS System Features
  • Sharp peak shapes and complete peak separation with temperature-ramped gas chromatography.
  • Full insight and control over the entire analytical workflow.
  • Widest dynamic range in EA-IRMS with multi-element analysis.
  • Lowest helium consumption and highest sample throughput.
  • Full system automation and lowest operating cost.
  • Flexibility for all applications with an easy-to-use modular system.
  • Unmatched robustness.
Especificações
IncludesFlash IRMS Elemental Analyzer, Delta Q IRMS, Conflo IV Universal Interface
Detector TypeThermal Conductivity Detector, IRMS
For Use With (Application)Geoscience, Food Safety, Environmental Science, Forensics, Petroleum, Exploration, Marine Science, Agronomy
TypeCNSOH IRMS System
Unit SizeEach