Inquire about OEM or Commercial Supply version of this product here.
Dynabeads™ Streptavidin Magnetic Beads
Invitrogen™

Dynabeads™ Streptavidin Magnetic Beads

Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
11206DDynabeads™ M-280 Streptavidin10 mL
65001Dynabeads™ MyOne™ Streptavidin C12 mL
65002Dynabeads™ MyOne™ Streptavidin C110 mL
65604DDynabeads™ MyOne™ Streptavidin T150 mL
65602Dynabeads™ MyOne™ Streptavidin T110 mL
65601Dynabeads™ MyOne™ Streptavidin T12 mL
65605DDynabeads™ Streptavidin for Target Enrichment2 mL
65606DDynabeads™ Streptavidin for Target Enrichment10 mL
65607DDynabeads™ Streptavidin for Target Enrichment50 mL
11205DDynabeads™ M-280 Streptavidin2 mL
60210Dynabeads™ M-280 Streptavidin100 mL
65305Dynabeads™ M-270 Streptavidin2 mL
65306Dynabeads™ M-270 Streptavidin10 mL
60101Dynabeads™ kilobaseBINDER™ Kit200 Isolations
65801DDynabeads™ Streptavidin Trial Kit4 x 1 mL
Catalog number 11206D
Price (USD)
2,350.65
Online exclusive
2,555.00
Save 204.35
Each
Product Type:
Dynabeads™ M-280 Streptavidin
Quantity:
10 mL
Price (USD)
2,350.65
Online exclusive
2,555.00
Save 204.35
Each

Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.

Dynabeads Streptavidin Magnetic Beads are compatible with high-throughput automation platforms, including KingFisher sample purification systems, ensuring seamless integration into new and existing workflows.

Key advantages of Dynabeads Streptavidin Magnetic Beads

  • ISO 13485–certified manufacturing—exceptional manufacturing control to ensure robust and consistent performance between production lots, meeting international quality standards for medical devices
  • Monosized, monodispersed magnetic beads—uniform particle size provides consistent and reproducible results across applications
  • Dual size options—available in 1-μm and 2.8-μm diameter specifications to accommodate diverse experimental requirements and application-specific needs
  • Multiple surface chemistry options—various surface functionalization chemistries available to optimize binding efficiency and compatibility for specific applications
  • Application-optimized design—size and surface chemistry combinations enable best-fit selection for targeted experimental protocols and assay requirements

Precision-engineered manufacturing delivers exceptional control over critical bead properties

Dynabeads Streptavidin products are based on magnetic polystyrene-based beads that are covalently coupled with recombinant streptavidin, creating a robust platform for target capture and isolation using biotinylated affinity binders. Dynabeads Streptavidin product manufacturing enables exceptional precision over three critical parameters: size distribution, magnetization properties, and surface chemical characteristics. This ensures consistent performance across batches and applications, providing the reliability necessary for reproducible experimental outcomes.

Optimized size selection enhances application-specific performance

The Dynabeads Streptavidin product line offers two distinct size options, each engineered to excel in specific experimental contexts. The 1-μm beads, available in the Dynabeads MyOne series, are ideally suited for applications requiring fast reaction kinetics, slow sedimentation rates, and high sensitivity-to-capacity ratios. These smaller beads provide enhanced surface area contact and reduced gravitational settling, making them particularly valuable for sensitive detection assays.

In contrast, the 2.8-μm beads, found in the Dynabeads M-270 and M-280 series, are perfect for applications demanding low background binding and reduced risk of aggregation in aggregation-prone applications. These larger beads offer even easier handling and faster magnetic separation compared to the Dynabeads MyOne series, facilitating streamlined workflow processing.

Tailored surface chemistry options address diverse experimental requirements

The Dynabeads Streptavidin product line includes two distinct surface chemistry configurations designed to optimize performance across varied experimental conditions. Hydrophobic surfaces are available in Dynabeads MyOne Streptavidin T1, Dynabeads Streptavidin for Target Enrichment, and Dynabeads M-280 Streptavidin products. These surfaces are BSA-blocked after streptavidin coupling making them ideal for high signal-to-noise applications such as immunoassays, target enrichment, and on-bead PCR for next-generation sequencing.

Alternatively, hydrophilic surfaces are featured in Dynabeads MyOne Streptavidin C1, Dynabeads Streptavidin for In Vitro Transcription, and Dynabeads M-270 Streptavidin products. These surfaces contain no BSA blocking and are particularly suitable for hydrophobic targets, harsh lysis conditions, sample preparation workflows for mass spectrometry, and applications demanding low or no nuclease activity, such as on-bead reverse transcription or in vitro transcription.

For researchers uncertain about which bead type best suits their application, a Dynabeads Streptavidin Trial Kit is available. It allows for side-by-side performance evaluation of different bead formulations under your specific experimental conditions, ensuring optimal selection for your research needs.

ISO 13485 certification ensures scalable quality and OEM support

Manufactured under ISO 13485 certification, Dynabeads Streptavidin products deliver outstanding batch-to-batch consistency. This robust quality assurance framework ensures that once customers develop an assay or workflow, they can rely on reproducible results across multiple experimental iterations. This consistency makes these versatile beads an efficient solution for applications spanning the entire spectrum from fundamental research to commercial manufacturing processes. For researchers developing commercial products or services, or those requiring larger bulk volumes, specialized support is available by emailing oemdynal@thermofisher.com.

For Research Use Only.
Specifications
Bead TypeMagnetic polystyrene-based beads covalently coupled with recombinant Streptavidin
Binding Property490-750 pmol free biotin/mg beads
Certifications/ComplianceISO9001 and ISO13485
Concentration10 mg/mL
Diameter (Metric)2.8 μm (CV < 5%)
For Use With (Application)Immunoassays, biopanning, etc, requiring high specificity, low background binding, and ease of handling
For Use With (Equipment)KingFisher™ Sample Purification System, DynaMag™ magnets
High-throughput CompatibilityHigh-throughput Compatible
Product LineDynabeads
Product TypeDynabeads™ M-280 Streptavidin
Quantity10 mL
Regulatory StatusRUO
Shelf Life36 months from date of manufacture
Shipping ConditionAmbient Temperature
Surface FunctionalityTosylactivated hydrophobic surface blocked with BSA
FormatBeads in Suspension
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
Dynabeads M-280 Streptavidin is supplied in PBS, pH 7.4 with 0.02% NaN3. Store at 2°-8°C.

Frequently asked questions (FAQs)

Can Dynabeads Streptavidin magnetic beads be boiled?

We do not recommend this as streptavidin becomes hydrophobic and aggregates during denaturation.

I am getting high background using Dynabeads M-280 Streptavidin magnetic beads. How can I prevent this from happening?

The background might be caused by nonspecific binding to the BSA on the bead surface. Alternatively, high background might be caused by nonspecific binding to streptavidin. Increasing either the pH or the salt concentration might help reduce the binding. Dynabeads M-270 Streptavidin magnetic beads might be a better alternative; these beads are not coated with BSA and are hydrophilic, as they are based upon carboxylic acid chemistry.

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

My Dynabeads M-280 Streptavidin magnetic beads were accidentally frozen. Can I still use them?

Yes, you can still use them. To test stability at low temperature in-house, the Dynabeads M-280 Streptavidin beads were placed at -20 degrees C for 24 hrs and then transferred to room temperature (15-25 degrees C) for 72 hrs. This freeze/thaw cycle was repeated 4 times and the beads were stored at 2-8 degrees C for 60 months. The biotin binding capacity was then tested and was seen to be within our specifications.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Which streptavidin-conjugated Dynabeads magnetic beads are the right beads for my application?

Which product to choose depends on the properties of your sample, the buffers and solutions applied, as well as the downstream application. In general, all Dynabeads Streptavidin beads can be used in applications involving biotinylated ligands; however, some beads may perform better than others in particular applications due to their characteristics. Dynabeads M-280 Streptavidin beads and Dynabeads MyOne Streptavidin T1 beads are commonly used for protein and nucleic acid applications. Dynabeads M-270 Streptavidin beads and MyOne Streptavidin C1 beads are preferred for nucleic acid diagnostics, specifically with samples that have a high concentration of chaotropic salts, immunoassays involving small biotinylated antigens, and in applications that are not compatible with BSA, as these beads are not blocked with BSA. Dynabeads MyOne Streptavidin beads offer increased binding capacity and slower sedimentation rate, making them ideal for automated applications and when larger amounts of a biotinylated compound or its specific target need to be isolated. Please see the selection guide here ( https://www.thermofisher.com/us/en/home/brands/product-brand/dynal/streptavidin-coupled-dynabeads.html?icid=fr-strep-1).

Citations & References (13)

Citations & References
Abstract
AID-induced decrease in topoisomerase 1 induces DNA structural alteration and DNA cleavage for class switch recombination.
Authors:Kobayashi M, Aida M, Nagaoka H, Begum NA, Kitawaki Y, Nakata M, Stanlie A, Doi T, Kato L, Okazaki IM, Shinkura R, Muramatsu M, Kinoshita K, Honjo T,
Journal:Proc Natl Acad Sci U S A
PubMed ID:20018730
'To initiate class switch recombination (CSR) activation-induced cytidine deaminase (AID) induces staggered nick cleavage in the S region, which lies 5'' to each Ig constant region gene and is rich in palindromic sequences. Topoisomerase 1 (Top1) controls the supercoiling of DNA by nicking, rotating, and religating one strand of DNA. ... More
Recommended mass spectrometry-based strategies to identify botulinum neurotoxin-containing samples.
Authors:Kalb SR, Baudys J, Wang D, Barr JR
Journal:
PubMed ID:25996606
'Botulinum neurotoxins (BoNTs) cause the disease called botulism, which can be lethal. BoNTs are proteins secreted by some species of clostridia and are known to cause paralysis by interfering with nerve impulse transmission. Although the human lethal dose of BoNT is not accurately known, it is estimated to be between ... More
Using phage and yeast display to select hundreds of monoclonal antibodies: application to antigen 85, a tuberculosis biomarker.
Authors:Ferrara F, Naranjo LA, Kumar S, Gaiotto T, Mukundan H, Swanson B, Bradbury AR
Journal:PLoS One
PubMed ID:23166701
'Current diagnostic methods for tuberculosis (TB), a major global health challenge that kills nearly two million people annually, are time-consuming and inadequate. During infection a number of bacterial molecules that play a role in the infective process are released and have been proposed as biomarkers for early TB diagnosis. Antigen ... More
PCR-free detection of genetically modified organisms using magnetic capture technology and fluorescence cross-correlation spectroscopy.
Authors:Zhou X, Xing D, Tang Y, Chen WR,
Journal:PLoS One
PubMed ID:19956680
'The safety of genetically modified organisms (GMOs) has attracted much attention recently. Polymerase chain reaction (PCR) amplification is a common method used in the identification of GMOs. However, a major disadvantage of PCR is the potential amplification of non-target DNA, causing false-positive identification. Thus, there remains a need for a ... More
PK assays for antibody-drug conjugates: case study with ado-trastuzumab emtansine.
Authors:Dere R, Yi JH, Lei C, Saad OM, Huang C, Li Y, Baudys J, Kaur S
Journal:Bioanalysis
PubMed ID:23641694
Antibody-drug conjugates (ADCs) combine the characteristics of large-molecule biologics and small-molecule drugs and are heterogeneous mixtures that can biotransform in vivo, resulting in additional complexity. ADC bioanalytical strategies require novel analytical methods, as well as existing large- and small-molecule methods. Because ADCs in late-stage clinical development are relatively new, regulatory ... More