3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water
3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water
3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water
Thermo Scientific Chemicals

3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water

CAS: 868272-85-9 | C12H18Cl4N4 | 360.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
11209025025 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 112090250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS868272-85-9
IUPAC Name[1,1'-biphenyl]-3,3',4,4'-tetramine tetrahydrochloride
Molecular FormulaC12H18Cl4N4
InChI KeyKJDSORYAHBAGPP-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<10 % (120°C)
Appearance (Color)Grey to brown-purple
Appearance (Form)Crystalline powder
Titration Argentometric>=96.0 % (on dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the water solubility of 3,3'-Diaminobenzidine tetrahydrochloride hydrate (Cat. No. 112090250)?

3,3'-Diaminobenzidine tetrahydrochloride hydrate is soluble in water at 50 mg/ml.