Interested in a bulk or custom version of this product? Request pricing.

CHI3L1 Recombinant Human Protein, His Tag

Catalog number: 11227H08H5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Chitinase 3-Like 1 (Cartilage Glycoprotein-39) (CHI3L1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications

This recombinant protein was expressed from a DNA sequence encoding the human CHI3L1 (NP_001267.2) (Met 1-Thr 383) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by the ability of the immobilized protein to support the adhesion of Fadu human squamous cell carcinoma cells (ATCC:HTB43). When cells are added to recombinant human CHI3L1 coated plates (1.5 µg/ml, 100 µl/well), approximately 65%-75% will adhere specifically after 30 minutes at 37°C.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Frozen
Gene Aliases: ASRT7, DKFZp686N19119, FLJ38139, GP39, HC-gp39, HCGP-3P, YKL40, YYL-40
Gene Symbol: CHI3L1
Molecular Weight: 42 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Family: Other Proteins
Protein Name: Chitinase 3-like 1 (cartilage glycoprotein-39)
Protein Subtype: Other Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 95% by SDS-PAGE
Research Category: Biomarker-Related Pathway(s), Immunology, Inflammation, Oncology, Signal Transduction
Species: Human
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols