1,5-Diaminonaphthalene, 97%
1,5-Diaminonaphthalene, 97%
1,5-Diaminonaphthalene, 97%
Thermo Scientific Chemicals

1,5-Diaminonaphthalene, 97%

CAS: 2243-62-1 | C10H10N2 | 158.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 112291000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2243-62-1
IUPAC Namenaphthalene-1,5-diamine
Molecular FormulaC10H10N2
InChI KeyKQSABULTKYLFEV-UHFFFAOYSA-N
SMILESNC1=CC=CC2=C(N)C=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
GC>=96.0 %
Appearance (Color)Grey to dark brown
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only