Interested in a bulk or custom version of this product? Request pricing.

SMYD3 Recombinant Human Protein, His Tag N-Flag

Catalog number: 11230H18H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

SET and MYND domain containing 3 (SMYD3), recombinant human protein is supplied in frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human SMYD3 isoform 1 (NP_001161212.1) (Met 1-Ser 428) fused to the flag tag at the N-terminus and the Histidine tag at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 25 mM Tris, 100 mM NaCl, 20% glycerol, 3 mM DTT, pH 8.0..

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Methylation & Demethylation Proteins
Research Category: Oncology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Frozen
Gene Aliases: FLJ21080, KMT3E, MGC104324, ZMYND1, ZNFN3A1, bA74P14.1, SMYD3
Gene ID: 64754
Gene Symbol: SMYD3
Molecular Weight: 51.5 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Form: Full Length, Recombinant
Protein Name: SET and MYND domain containing 3
Protein Tag or Fusion: N-Flag C-His
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols