Interested in a bulk or custom version of this product? Request pricing

gp120 (fragment of env) Recombinant HIV Protein, His Tag, Active

Catalog number: 11233V08H5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

HIV-1 glycoprotein 120 (gp120, fragment of env), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the HIV-1 surface glycoprotein gp120 (Ala 29-Glu 503), which is cleaved from the full length envelope glycoprotein gp160, fused to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.

Activity: Measured by its binding ability in a functional ELISA. Immobilized human CD4-His at 2 µg/well can bind biotinylated HIV-1 GP120 with a linear range of 0.078-1.25 µg/ml.

Formulation: Lyophilized in PBS, pH7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: gp120, SU, Surface glycoprotein 120
Gene ID: 155971
Gene Symbol: gp120
Molecular Weight: 54.6 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Form: Recombinant
Protein Name: Surface glycoprotein 120
Protein Subtype: Viral Proteins
Protein Tag or Fusion: His Tag
Purity: ≥ 95% by SDS-PAGE
Research Category: Immunology, Virology
Species: HIV
Validated Application: Assay Standard

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols