1,3-Diaminopropane, 99%
1,3-Diaminopropane, 99%
1,3-Diaminopropane, 99%
Thermo Scientific Chemicals

1,3-Diaminopropane, 99%

CAS: 109-76-2 | C3H10N2 | 74.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 112350500
Price (EUR)
21,65
Online exclusive
23,50
Save 1,85 (8%)
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
21,65
Online exclusive
23,50
Save 1,85 (8%)
Each
Chemical Identifiers
CAS109-76-2
IUPAC Namepropane-1,3-diamine
Molecular FormulaC3H10N2
InChI KeyXFNJVJPLKCPIBV-UHFFFAOYSA-N
SMILESNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Titration with HClO4>=98.5 %
GC>=98.5 %
Water=<0.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only