Interested in a bulk or custom version of this product? Request pricing.

CRP Recombinant Human Protein, His Tag

Catalog number: 11250H08H250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

C-Reactive Protein Pentraxin-Related (CRP) recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding human CRP (NP_000558.2) (Met 1-Pro 224) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to bind human CD32a (Cat. No.10374-H08B) in functional ELISA.

Formulation: Lyophilized in 20 mM Tris, 0.2 M NaCl, 5 mM CaCl2, pH 8.0.

Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: Mammalian
Form: Lyophilized
Gene Aliases: RP11-419N10.4, MGC149895, MGC88244, PTX1
Gene ID: 1401
Gene Symbol: CRP
Molecular Weight: 24.7 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: C-reactive protein, pentraxin-related
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Research Category: Biomarker-Related Pathway(s), Cardiovascular Research
Species: Human
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols