Dibenzylamine, 98%
Dibenzylamine, 98%
Dibenzylamine, 98%
Thermo Scientific Chemicals

Dibenzylamine, 98%

CAS: 103-49-1 | C14H15N | 197.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 112610010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-49-1
IUPAC Namedibenzylamine
Molecular FormulaC14H15N
InChI KeyBWLUMTFWVZZZND-UHFFFAOYSA-N
SMILESC(NCC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5740 to 1.5760 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only