AmnioMAX™-II Complete Medium
Gibco™

AmnioMAX™-II Complete Medium

Green features
Fully-supplemented medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures
Have Questions?
Catalog NumberQuantity
11269016100 mL
Catalog number 11269016
Price (USD)
150.00
Each
Quantity:
100 mL
Price (USD)
150.00
Each

Gibco™ AmnioMAX™ - II Complete Medium is a fully-supplemented medium developed for the short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures.

Gibco™ AmnioMAX™ - II Complete Medium features:

  • Ready-to-use format
  • Quality and performance testing
  • Unique, optimized formulation

Ready-to-use format

Gibco™ AmnioMAX™ - II Complete Medium is a frozen, 1X medium, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2–8°C for up to ten days.

Quality and performance testing

Every lot of Gibco™ AmnioMAX™ - II Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Pooled primary human amniotic fluid samples are cultured for six days in Gibco™ AmnioMAX™ - II Complete Medium before measuring the total number of colonies and total number of mitotic colonies. In addition, each lot is tested for pH and osmolality and must pass a sterility test prior to lot release.

Unique, optimized formulation

Gibco™ AmnioMAX™ - II Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth. This optimized medium also has an enhanced buffering system that provides greater pH stability during culture manipulations.

Product Use

For in vitro diagnostic use.

cGMP Manufacturing and Quality System

Gibco™ AmnioMAX™ - II Complete Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

WARNING: Cancer – www.P65Warnings.ca.gov
In vitro diagnostic
Specifications
Cell TypeAmniocyte Cells
Concentration1 X
Green FeaturesACT Ecolabel (Silver)
Included AntibioticsGentamicin
Product LineAmnioMAX
Product TypeComplete Medium
Quantity100 mL
Shelf Life18 Months
Shipping ConditionDry Ice
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesGlutamine
Unit SizeEach
Contents & Storage
Storage conditions: -5 to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 18 months from date of manufacture if frozen. If stored at 4°C (protected from light): use within 7–10 days once thawed and opened. Extended shelf-life of 50 days if thawed and unopened.

Frequently asked questions (FAQs)

Can I supplement AmnioMax-II Complete Medium (Cat. No. 11269016) with Fungizone?

No. We do not recommend supplementing AmnioMAX-II Complete Medium (Cat. No. 1129016) with Fungizone, as it can be detrimental to amniocyte growth.  

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (3)

Citations & References
Abstract
Induced pluripotent stem cells offer new approach to therapy in thalassemia and sickle cell anemia and option in prenatal diagnosis in genetic diseases.
Authors:Ye L, Chang JC, Lin C, Sun X, Yu J, Kan YW,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19482945
'The innovation of reprogramming somatic cells to induced pluripotent stem cells provides a possible new approach to treat beta-thalassemia and other genetic diseases such as sickle cell anemia. Induced pluripotent stem (iPS) cells can be made from these patients'' somatic cells and the mutation in the beta-globin gene corrected by ... More
Specific tumour-associated methylation in normal human term placenta and first-trimester cytotrophoblasts.
Authors:Novakovic B, Rakyan V, Ng HK, Manuelpillai U, Dewi C, Wong NC, Morley R, Down T, Beck S, Craig JM, Saffery R,
Journal:Mol Hum Reprod
PubMed ID:18708652
'Human placentation displays many similarities with tumourigenesis, including rapid cell division, migration and invasion, overlapping gene expression profiles and escape from immune detection. Recent data have identified promoter methylation in the Ras association factor and adenomatous polyposis coli tumour suppressor genes as part of this process. However, the extent of ... More
Placenta-specific methylation of the vitamin D 24-hydroxylase gene: implications for feedback autoregulation of active vitamin D levels at the fetomaternal interface.
Authors:Novakovic B, Sibson M, Ng HK, Manuelpillai U, Rakyan V, Down T, Beck S, Fournier T, Evain-Brion D, Dimitriadis E, Craig JM, Morley R, Saffery R,
Journal:J Biol Chem
PubMed ID:19237542
'Plasma concentrations of biologically active vitamin D (1,25-(OH)(2)D) are tightly controlled via feedback regulation of renal 1alpha-hydroxylase (CYP27B1; positive) and 24-hydroxylase (CYP24A1; catabolic) enzymes. In pregnancy, this regulation is uncoupled, and 1,25-(OH)(2)D levels are significantly elevated, suggesting a role in pregnancy progression. Epigenetic regulation of CYP27B1 and CYP24A1 has previously ... More