1,10-Dibromodecane, 97%
1,10-Dibromodecane, 97%
1,10-Dibromodecane, 97%
Thermo Scientific Chemicals

1,10-Dibromodecane, 97%

CAS: 4101-68-2 | C10H20Br2 | 300.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 112780250
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.30
Save 9.65
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.30
Save 9.65
Each
Chemical Identifiers
CAS4101-68-2
IUPAC Name1,10-dibromodecane
Molecular FormulaC10H20Br2
InChI KeyGTQHJCOHNAFHRE-UHFFFAOYSA-N
SMILESBrCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Fused solid or liquid
Infrared spectrumConforms
Melting point25°C to 30°C
GC>=96.0 %
Appearance (Color)White to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only